accessibility menu, dialog, popup

UserName

H3COOH3CH2NSOONH

CAS RN 52298-44-9

IUPAC Name: 4-amino-2,5-dimethoxy-N-phenylbenzene-1-sulfonamide
Synonyms: 4-amino-2,5-dimethoxy-N-phenylbenzenesulfonamide
Molecular Weight (g/mol): 308.35
Molecular Formula: C14H16N2O4S
InChi Key: NFYQAEPHDGXJSY-UHFFFAOYSA-N
SMILES: COC1=CC(=C(OC)C=C1N)S(=O)(=O)NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results