missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,5-dimethoxy-N-phenylbenzenesulfonamide 98.0+%, TCI America™

Catalog No. A198725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A198725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A198725G Supplier TCI America Supplier No. A198725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52298-44-9
Molecular Formula C14H16N2O4S
Molecular Weight (g/mol) 308.352
MDL Number MFCD00025753
InChI Key NFYQAEPHDGXJSY-UHFFFAOYSA-N
Synonym 4-amino-2,5-dimethoxy-n-phenylbenzenesulphonamide, unii-54s41p93bi, 2,5-dimethoxyaniline-4-sulfonanilide, benzenesulfonamide, 4-amino-2,5-dimethoxy-n-phenyl, 2,5-dimethoxyaniline-4-sulfoanilide, acmc-1akg1, 4-amino-2,5-dimethoxy-n-phenylbenzolsulfonamid, dsstox_cid_24686, dsstox_rid_80397, dsstox_gsid_44686
PubChem CID 104143
IUPAC Name 4-amino-2,5-dimethoxy-N-phenylbenzenesulfonamide
SMILES COC1=CC(=C(C=C1N)OC)S(=O)(=O)NC2=CC=CC=C2

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 25 g
Formula Weight 308.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,5-dimethoxy-N-phenylbenzenesulfonamide
EINECSNumber (3)-2093&(3)-3163
TSCA Yes