CAS RN 52193-54-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Promotions Available
Pyridinium trifluoromethanesulfonate, 97%
Pyridinium trifluoromethanesulfonate, 97%
CAS: 52193-54-1 MDL Number: MFCD00274656 Synonym: pyridinium trifluoromethanesulfonate,pyridinium triflate,pyridin-1-ium trifluoromethanesulfonate,pyridium triflate,acmc-209ky5,pyridinium trifluoromethanesulphonate,pyridin-1-ium; trifluoromethanesulfonate
2
Pyridinium Trifluoromethanesulfonate 98.0+%, TCI America™
CAS: 52193-54-1 Molecular Formula: C6H6F3NO3S Synonym: pyridinium trifluoromethanesulfonate,pyridinium triflate,pyridin-1-ium trifluoromethanesulfonate,pyridium triflate,acmc-209ky5,pyridinium trifluoromethanesulphonate,pyridin-1-ium; trifluoromethanesulfonate