missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. P16275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16275G 5 g
P162725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16275G Supplier TCI America Supplier No. P16275G

Chemical Identifiers

CAS 52193-54-1
Molecular Formula C6H6F3NO3S
Synonym pyridinium trifluoromethanesulfonate, pyridinium triflate, pyridin-1-ium trifluoromethanesulfonate, pyridium triflate, acmc-209ky5, pyridinium trifluoromethanesulphonate, pyridin-1-ium; trifluoromethanesulfonate

Specifications

Melting Point 224°C
Color White
Quantity 5 g
Formula Weight 229.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium Trifluoromethanesulfonate