accessibility menu, dialog, popup

UserName

CAS RN 5211-44-9

IUPAC Name: 2,3,5,6-tetrafluoro-4-sulfanylbenzoic acid
Synonyms: 2,3,5,6-tetrafluoro-4-sulfanylbenzoic acid
Molecular Weight (g/mol): 226.14
Molecular Formula: C7H2F4O2S
InChi Key: USFMEWZQIHKRDP-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(F)=C(S)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results