accessibility menu, dialog, popup

UserName

HClHON

CAS RN 52-49-3

IUPAC Name: hydrogen 1-cyclohexyl-1-phenyl-3-(piperidin-1-yl)propan-1-ol chloride
Synonyms: hydrogen trihexyphenidyl chloride
Molecular Weight (g/mol): 337.93
Molecular Formula: C20H32ClNO
InChi Key: QDWJJTJNXAKQKD-UHFFFAOYNA-N
SMILES: [H+].[Cl-].OC(CCN1CCCCC1)(C1CCCCC1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results