missing translation for 'onlineSavingsMsg'
Learn More

Trihexyphenidyl Hydrochloride 98.0+%, TCI America™

Catalog No. T33035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33035G 5 g
T330325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33035G Supplier TCI America Supplier No. T33035G
Only null left

Chemical Identifiers

CAS 52-49-3
Molecular Formula C20H32ClNO
Molecular Weight (g/mol) 337.93
MDL Number MFCD00058212
InChI Key QDWJJTJNXAKQKD-UHFFFAOYNA-N
Synonym 1-Cyclohexyl-1-phenyl-3-(1-piperidyl)-1-propanol Hydrochloride, alpha-Cyclohexyl-alpha-phenyl-1-piperidinepropanol Hydrochloride, Benzhexol Hydrochloride
PubChem CID 66007
IUPAC Name hydrogen 1-cyclohexyl-1-phenyl-3-(piperidin-1-yl)propan-1-ol chloride
SMILES [H+].[Cl-].OC(CCN1CCCCC1)(C1CCCCC1)C1=CC=CC=C1

Specifications

Melting Point 247°C
Color White
Quantity 5 g
Formula Weight 337.93
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trihexyphenidyl Hydrochloride
RTECSNumber TN2625000
TSCA No