accessibility menu, dialog, popup

UserName

HOOOHOHO

CAS RN 518-44-5

IUPAC Name: 2-(3,6-dihydroxy-9H-xanthen-9-yl)benzoic acid
Synonyms: fluorescin
Molecular Weight (g/mol): 334.33
Molecular Formula: C20H14O5
InChi Key: MURGITYSBWUQTI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C1C2=CC=C(O)C=C2OC2=CC(O)=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results