accessibility menu, dialog, popup

UserName

H3CONHH2NHCl

CAS RN 51721-68-7

IUPAC Name: 4-methoxybenzene-1-carboximidamide hydrochloride
Synonyms: 4-methoxybenzenecarboximidamide hydrochloride
Molecular Weight (g/mol): 186.64
Molecular Formula: C8H11ClN2O
InChi Key: AJOSDIDPIBJFAI-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=C(C=C1)C(N)=N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results