missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzamidine Hydrochloride 98.0+%, TCI America™

Catalog No. M26125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26125G 5 g
M26121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26125G Supplier TCI America Supplier No. M26125G

Chemical Identifiers

CAS 51721-68-7
Molecular Formula C8H11ClN2O
Molecular Weight (g/mol) 186.639
MDL Number MFCD00466114
InChI Key AJOSDIDPIBJFAI-UHFFFAOYSA-N
PubChem CID 12326219
IUPAC Name 4-methoxybenzenecarboximidamide;hydrochloride
SMILES COC1=CC=C(C=C1)C(=N)N.Cl

Specifications

Melting Point 221°C
Color White
Quantity 5 g
Formula Weight 186.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxybenzamidine Hydrochloride