accessibility menu, dialog, popup

UserName

CAS RN 51698-33-0

IUPAC Name: disodium 2-chloro-5-(4-chloro-3-sulfonatobenzenesulfonyl)benzene-1-sulfonate
Synonyms: disodium 2-chloro-5-(4-chloro-3-sulfonatobenzenesulfonyl)benzenesulfonate
Molecular Weight (g/mol): 491.23
Molecular Formula: C12H6Cl2Na2O8S3
InChi Key: KKEBUZUONXHUNE-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=C(Cl)C=CC(=C1)S(=O)(=O)C1=CC(=C(Cl)C=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results