accessibility menu, dialog, popup

UserName

ClOOHCl

CAS RN 51656-68-9

IUPAC Name: 3-(2,6-dichlorophenyl)propanoic acid
Synonyms: 3-(2,6-dichlorophenyl)propanoic acid
Molecular Weight (g/mol): 219.06
Molecular Formula: C9H8Cl2O2
InChi Key: IDEOVPXKPDUXTP-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=C(Cl)C=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results