missing translation for 'onlineSavingsMsg'
Learn More

3-(2,6-Dichlorophenyl)propionic acid, 96%

Catalog No. AAH3321014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3321014 25 g
AAH3321003 1 g
AAH3321006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3321014 Supplier Thermo Scientific Chemicals Supplier No. H3321014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51656-68-9 | C9H8Cl2O2 | 219.061 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51656-68-9
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.061
MDL Number MFCD01310807
InChI Key IDEOVPXKPDUXTP-UHFFFAOYSA-N
PubChem CID 2758192
IUPAC Name 3-(2,6-dichlorophenyl)propanoic acid
SMILES C1=CC(=C(C(=C1)Cl)CCC(=O)O)Cl

Specifications

Melting Point 97°C to 99°C
Quantity 25 g
Synonym 3-2,6-dichlorophenyl propanoic acid, 3-2,6-dichlorophenyl propionic acid, 3-2,6-dichlorophenyl propanoicacid, benzenepropanoic acid,2,6-dichloro, benzenepropanoic acid, 2,6-dichloro, 3-2,6-bis chloranyl phenyl propanoic acid
Formula Weight 219.07
Percent Purity 96%
Chemical Name or Material 3-(2,6-Dichlorophenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only