accessibility menu, dialog, popup

UserName

CAS RN 5138-90-9

IUPAC Name: sodium 4-chlorobenzene-1-sulfonate
Synonyms: sodium 4-chlorobenzenesulfonate
Molecular Weight (g/mol): 214.59
Molecular Formula: C6H4ClNaO3S
InChi Key: XLKHCFJHGIAKFX-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)