missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Chlorobenzenesulfonate 98.0+%, TCI America™

Catalog No. C0127500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0127500G 500 g
C012725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0127500G Supplier TCI America Supplier No. C0127500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5138-90-9
Molecular Formula C6H4ClNaO3S
Molecular Weight (g/mol) 214.59
MDL Number MFCD00014731
InChI Key XLKHCFJHGIAKFX-UHFFFAOYSA-M
Synonym 4-Chlorobenzenesulfonic Acid Sodium Salt
PubChem CID 517331
IUPAC Name sodium 4-chlorobenzene-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Color White
Quantity 500 g
Formula Weight 214.59
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Chlorobenzenesulfonate
RTECSNumber DB5620000
TSCA Yes