accessibility menu, dialog, popup

UserName

H3CPOHOHO

CAS RN 5137-70-2

IUPAC Name: dodecylphosphonic acid
Synonyms: phosphonic acid, dodecyl-
Molecular Weight (g/mol): 250.32
Molecular Formula: C12H27O3P
InChi Key: SVMUEEINWGBIPD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCP(O)(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)