accessibility menu, dialog, popup

UserName

H3CCH3I

CAS RN 513-48-4

IUPAC Name: 2-iodobutane
Synonyms: 2-iodobutane
Molecular Weight (g/mol): 184.02
Molecular Formula: C4H9I
InChi Key: IQRUSQUYPCHEKN-UHFFFAOYNA-N
SMILES: CCC(C)I
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)