missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobutane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. B109325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B109325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B109325G Supplier TCI America Supplier No. B109325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 513-48-4
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001072
InChI Key IQRUSQUYPCHEKN-UHFFFAOYNA-N
Synonym sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide
PubChem CID 10559
IUPAC Name 2-iodobutane
SMILES CCC(C)I

Specifications

Boiling Point 120°C
Quantity 25 g
UN Number 2390
Formula Weight 184.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodobutane (stabilized with Copper chip)
RTECSNumber EK4410000
TSCA Yes