accessibility menu, dialog, popup

UserName

HONHHNHNNHOOOH

CAS RN 51094-17-8

IUPAC Name: 4-[7,12-bis(4-hydroxyphenyl)-17-(4-oxocyclohexa-2,5-dien-1-ylidene)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),3,5,7,9,11,13,15,18-nonaen-2-ylidene]cyclohexa-2,5-dien-1-one
Synonyms: 4-[7,12-bis(4-hydroxyphenyl)-17-(4-oxocyclohexa-2,5-dien-1-ylidene)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),3,5,7,9,11,13,15,18-nonaen-2-ylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 678.75
Molecular Formula: C44H30N4O4
InChi Key: OXBXIINNAXASCM-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=C2NC(C=C2)=C(C2=CC=C(N2)C(C2=CC=C(N2)C(C2=CC=C1N2)=C1C=CC(=O)C=C1)=C1C=CC(=O)C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results