missing translation for 'onlineSavingsMsg'
Learn More

5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin 95.0+%, TCI America™

Catalog No. T1497100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1497100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1497100MG Supplier TCI America Supplier No. T1497100MG
Only null left

Chemical Identifiers

CAS 51094-17-8
Molecular Formula C44H30N4O4
Molecular Weight (g/mol) 678.75
MDL Number MFCD00191501
InChI Key OXBXIINNAXASCM-UHFFFAOYSA-N
Synonym 5,10,15,20-Tetrakis(4-hydroxyphenyl)-21H,23H-porphine
PubChem CID 6847207
IUPAC Name 4-[7,12-bis(4-hydroxyphenyl)-17-(4-oxocyclohexa-2,5-dien-1-ylidene)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),3,5,7,9,11,13,15,18-nonaen-2-ylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC=C(C=C1)C1=C2NC(C=C2)=C(C2=CC=C(N2)C(C2=CC=C(N2)C(C2=CC=C1N2)=C1C=CC(=O)C=C1)=C1C=CC(=O)C=C1)C1=CC=C(O)C=C1

Specifications

Color Black-Purple
Quantity 100 mg
Formula Weight 678.75
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin