accessibility menu, dialog, popup

UserName

CH3OHOF

CAS RN 5104-49-4

IUPAC Name: 2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
Synonyms: flurbiprofen flubiprofen froben strefen shire brand of flurbiprofen ratiopharm brand of flurbiprofen novo-flurprofen 氟比洛芬 klosterfrau brand of flurbiprofen apotex brand of flurbiprofen
Molecular Weight (g/mol): 244.27
Molecular Formula: C15H13FO2
InChi Key: SYTBZMRGLBWNTM-UHFFFAOYNA-N
SMILES: CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)