missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-alpha-methyl-4-biphenylacetic acid, 99%

Catalog No. AAB2260314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2260314 25 g
AAB2260303 1 g
AAB2260306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2260314 Supplier Thermo Scientific Chemicals Supplier No. B2260314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5104-49-4 | C15H13FO2 | 244.27 g/mol

A cyclooxygenase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5104-49-4
Molecular Formula C15H13FO2
Molecular Weight (g/mol) 244.27
MDL Number MFCD00079303
InChI Key SYTBZMRGLBWNTM-UHFFFAOYNA-N
Synonym flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
PubChem CID 3394
ChEBI CHEBI:5130
SMILES CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1

Specifications

Melting Point 112°C to 114°C
Odor Odorless
Quantity 25 g
UN Number UN2811
Merck Index 14,4199
IUPAC Name 2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
Formula Weight 244.27
Percent Purity 99%
Chemical Name or Material 2-Fluoro-alpha-methyl-4-biphenylacetic acid
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 225-827-6
RTECSNumber DU8341000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only