missing translation for 'onlineSavingsMsg'
Learn More

Flurbiprofen 98.0+%, TCI America™

Catalog No. F03715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03715G 5 g
F037125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03715G Supplier TCI America Supplier No. F03715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5104-49-4
Molecular Formula C15H13FO2
Molecular Weight (g/mol) 244.27
MDL Number MFCD00079303
InChI Key SYTBZMRGLBWNTM-UHFFFAOYNA-N
Synonym flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
PubChem CID 3394
ChEBI CHEBI:5130
IUPAC Name 2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
SMILES CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1

Specifications

Melting Point 113°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 244.27
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Flurbiprofen
RTECSNumber DU8341000
TSCA No