missing translation for 'onlineSavingsMsg'
Learn More

Flurbiprofen Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017822
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017822 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017822 Supplier Merck Emd Millipore Supplier No. PHR1499500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Flurbiprofen is a non-selective, non-steroidal anti-inflammatory drug (NSAID) that is widely used against rheumatoid arthritis. It is also effective against vernal keratoconjunctivitis, post-operative ocular inflammation, herpetic stromal keratitis, excimer laser photorefractive keratectomy and ocular gingivitis. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 5104-49-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C6H5C6H3(F)CH(CH3)CO2 H
MDL Number MFCD00079303
Melting Point 110°C to 112°C
Synonym Flurbiprofen; (+/-)-2-Fluoro-alpha-methyl-4-biphenylacetic acid; L-790,330
Type Certified Reference Material
Quantity 500 mg
Formula Weight 244.26
Grade Certified Reference Material