accessibility menu, dialog, popup

UserName

CH3ONHSHN

CAS RN 51-52-5

IUPAC Name: 6-propyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
Synonyms: propylthiouracil|6-N-propylthiouracil|6-N-propyl-2-thiouracil|propyl thiouracil|propythiouracil|propyl-thyracil|propilthiouracil|propylthiouracile|procasil|propacil
Molecular Weight (g/mol): 170.23
Molecular Formula: C7H10N2OS
InChi Key: KNAHARQHSZJURB-UHFFFAOYSA-N
SMILES: CCCC1=CC(=O)NC(=S)N1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)