missing translation for 'onlineSavingsMsg'
Learn More

6-Propyl-2-thiouracil 99.0+%, TCI America™

Catalog No. P053325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P053325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P053325G Supplier TCI America Supplier No. P053325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51-52-5
Molecular Formula C7H10N2OS
Molecular Weight (g/mol) 170.23
MDL Number MFCD00006041
InChI Key KNAHARQHSZJURB-UHFFFAOYSA-N
Synonym propylthiouracil, 6-propyl-2-thiouracil, propacil, prothiurone, prothyran, procasil, propycil, prothiucil, protiural, thiuragyl
PubChem CID 657298
ChEBI CHEBI:8502
IUPAC Name 6-propyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
SMILES CCCC1=CC(=O)NC(=S)N1

Specifications

Melting Point 219°C
Color White
pH 6.0 to 7.0 (5g/L 20°C)
Quantity 25 g
Formula Weight 170.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Propyl-2-thiouracil
EINECSNumber (5)-0936&(5)-3810
RTECSNumber YR1400000
TSCA Yes