missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-mercapto-6-propylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog No. AC121310250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121310250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC121310250 Supplier Thermo Scientific Chemicals Supplier No. 121310250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-52-5
Molecular Formula C7H10N2OS
Molecular Weight (g/mol) 170.23
MDL Number MFCD00006041
InChI Key KNAHARQHSZJURB-UHFFFAOYSA-N
Synonym propylthiouracil, 6-propyl-2-thiouracil, propacil, prothiurone, prothyran, procasil, propycil, prothiucil, protiural, thiuragyl
PubChem CID 657298
ChEBI CHEBI:8502
IUPAC Name 6-propyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
SMILES CCCC1=CC(=O)NC(=S)N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 218°C to 221°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Merck Index 15, 7980
Formula Weight 170.24
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 4-Hydroxy-2-mercapto-6-propylpyrimidine, 98%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-103-2
RTECSNumber YR1400000
TSCA TSCA

RUO – Research Use Only