accessibility menu, dialog, popup

UserName

CAS RN 5082-74-6

IUPAC Name: (pyrrolidin-3-yl)methanol
Synonyms: pyrrolidin-3-ylmethanol
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: QOTUIIJRVXKSJU-UHFFFAOYNA-N
SMILES: OCC1CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)