missing translation for 'onlineSavingsMsg'
Learn More

DL-beta-Prolinol, 97+%, Thermo Scientific Chemicals

Catalog No. AAH5722503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5722503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5722503 Supplier Thermo Scientific Chemicals Supplier No. H5722503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5082-74-6 | Molecular Formula: C5H11NO | Molecular Weight: 101.15 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.

Chemical Identifiers

CAS 5082-74-6
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD07773080
InChI Key QOTUIIJRVXKSJU-UHFFFAOYNA-N
Synonym 3-hydroxymethylpyrrolidine, 3-pyrrolidinemethanol, pyrrolidin-3-yl-methanol, pyrrolidine-3-yl-methanol, pyrrolidin-3-yl methanol, 3-pyrrolidinylmethanol, dl-beta-prolinol, rs-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine, 3-pyrrolidinemethanol, 3s
PubChem CID 13400657
IUPAC Name (pyrrolidin-3-yl)methanol
SMILES OCC1CCNC1

Specifications

Density 0.979
Boiling Point 165°C to 171°C
Quantity 1 g
Sensitivity Hygroscopic
Beilstein 1420904
Solubility Information Slightly soluble in water
Formula Weight 101.15
Percent Purity ≥97%
Chemical Name or Material DL-beta-Prolinol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only