accessibility menu, dialog, popup

UserName

CAS RN 50709-36-9

IUPAC Name: hydrogen (2,6-dichlorophenyl)hydrazine chloride
Synonyms: hydrogen (2,6-dichlorophenyl)hydrazine chloride
Molecular Weight (g/mol): 213.49
Molecular Formula: C6H7Cl3N2
InChi Key: CQNIYLLTIOPFCJ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=C(Cl)C=CC=C1Cl
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)