accessibility menu, dialog, popup

UserName

NNOOH

CAS RN 50681-25-9

IUPAC Name: pyridazine-4-carboxylic acid
Synonyms: pyridazine-4-carboxylic acid
Molecular Weight (g/mol): 124.1
Molecular Formula: C5H4N2O2
InChi Key: JUSIWJONLKBPDU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=NC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results