missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridazinecarboxylic acid, 95%

Catalog No. AC399630050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399630050 5 g
AC399630010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399630050 Supplier Thermo Scientific Chemicals Supplier No. 399630050

CAS: 50681-25-9 | C5H4N2O2 | 124.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50681-25-9
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.1
InChI Key JUSIWJONLKBPDU-UHFFFAOYSA-N
Synonym 4-pyridazinecarboxylic acid, 4-carboxypyridazine, 4-pyridazinecarboxylicacid, pubchem9501, acmc-1alr8, pyridazin-4-carboxylic acid, 4-pyridazine-carboxylic acid, ksc269i2f, pyridazin-4-yl formate, 1,2-diazine-4-carboxylic acid
PubChem CID 2761046
IUPAC Name pyridazine-4-carboxylic acid
SMILES C1=CN=NC=C1C(=O)O

Specifications

Melting Point 244.2°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 124.1
Percent Purity 95%
Chemical Name or Material 4-Pyridazinecarboxylic acid
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only