accessibility menu, dialog, popup

UserName

NH2NH2HNOHNOO

CAS RN 506-93-4

IUPAC Name: guanidine; nitric acid
Synonyms: guanidine nitrate
Molecular Weight (g/mol): 122.08
Molecular Formula: CH6N4O3
InChi Key: CNUNWZZSUJPAHX-UHFFFAOYSA-N
SMILES: NC(N)=N.O[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)