missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Nitrate 98.0+%, TCI America™

Catalog No. G016425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G016425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G016425G Supplier TCI America Supplier No. G016425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 506-93-4
Molecular Formula CH6N4O3
Molecular Weight (g/mol) 122.084
MDL Number MFCD00013028
InChI Key CNUNWZZSUJPAHX-UHFFFAOYSA-N
Synonym guanidine nitrate, guanidinium nitrate, guanidine; nitric acid, guanidine, mononitrate, guanidine, nitrate, guanidine mononitrate, guanidine nitrate 1:1, guanidinenitrate, pubchem21372, acmc-209kpa
PubChem CID 10481
IUPAC Name guanidine;nitric acid
SMILES C(=N)(N)N.[N+](=O)(O)[O-]

Specifications

Melting Point 214°C
Color White
Quantity 25 g
UN Number 1467
Formula Weight 122.08
Percent Purity ≥98.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Nitrate
EINECSNumber (1)-0394&(2)-1773
RTECSNumber MF4350000
TSCA Yes