accessibility menu, dialog, popup

UserName

CAS RN 5058-42-4

IUPAC Name: iron(3+) triethanolate
Synonyms: iron(3+) tris(ethoxide)
Molecular Weight (g/mol): 191.03
Molecular Formula: C6H15FeO3
InChi Key: JTPUGUWXHGEEHW-UHFFFAOYSA-N
SMILES: [Fe+3].CC[O-].CC[O-].CC[O-]
Boiling Point 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)