accessibility menu, dialog, popup

UserName

CAS RN 505-73-7

HOOSSOHO

CAS RN 505-73-7

IUPAC Name: 2-[(carboxymethyl)disulfanyl]acetic acid
Synonyms: dithiodiglycolic acid
Molecular Weight (g/mol): 182.21
Molecular Formula: C4H6O4S2
InChi Key: DLLMHEDYJQACRM-UHFFFAOYSA-N
SMILES: OC(=O)CSSCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results