missing translation for 'onlineSavingsMsg'
Learn More

Dithiodiglycolic Acid 96.0+%, TCI America™

Catalog No. D094525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D094525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D094525G Supplier TCI America Supplier No. D094525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 505-73-7
Molecular Formula C4H6O4S2
Molecular Weight (g/mol) 182.208
MDL Number MFCD00039607
InChI Key DLLMHEDYJQACRM-UHFFFAOYSA-N
Synonym dithiodiacetic acid, acetic acid, 2,2'-dithiobis, dithioglycolic acid, thioglycolate disulfide, acetic acid, dithiodi, 2,2'-dithiodiacetic acid, unii-7zz0lal4ti, 7zz0lal4ti, 2-carboxymethyldisulfanyl acetic acid, carboxymethyl disulfanyl acetic acid
PubChem CID 68164
IUPAC Name 2-(carboxymethyldisulfanyl)acetic acid
SMILES C(C(=O)O)SSCC(=O)O

Specifications

Melting Point 107°C
Color White
Quantity 25 g
Formula Weight 182.21
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dithiodiglycolic Acid
EINECSNumber (2)-1365
TSCA Yes