accessibility menu, dialog, popup

UserName

CAS RN 5034-74-2

IUPAC Name: 5-bromo-2-hydroxy-3-methoxybenzaldehyde
Synonyms: 5-bromo-2-hydroxy-3-methoxybenzaldehyde
Molecular Weight (g/mol): 231.05
Molecular Formula: C8H7BrO3
InChi Key: MMFKBTPDEVLIOR-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=CC(C=O)=C1O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)