missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxy-3-methoxybenzaldehyde, 97%, Thermo Scientific™

Catalog No. BTB03758EE
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB03758EE 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB03758EE Supplier Thermo Scientific Chemicals Supplier No. BTB03758EE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5034-74-2
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD00016593
InChI Key MMFKBTPDEVLIOR-UHFFFAOYSA-N
PubChem CID 262238
IUPAC Name 5-bromo-2-hydroxy-3-methoxybenzaldehyde
SMILES COC1=C(C(=CC(=C1)Br)C=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3OC6H2(OH)(Br)CH(O)
Quantity 50 g
Synonym 5-bromo-3-methoxysalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy-3-methoxy, 5-bromo-2-vanillin, acmc-20am5q, 3-methoxy-5-bromosalicylaldehyde, 5bromo-2-hydroxy-3-methoxy-benzaldehyde, 5-bromo-2-hydroxy-3-methoxy-benzaldehyde, benzaldehyde,5-bromo-2-hydroxy-3-methoxy, 5-bromo-2-hydroxy-3-methoxybenzaldehyde
Formula Weight 231.04
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-hydroxy-3-methoxybenzaldehyde
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.