accessibility menu, dialog, popup

UserName

CAS RN 5005-36-7

IUPAC Name: 2-phenyl-2-(pyridin-2-yl)acetonitrile
Synonyms: phenyl-(2-pyridyl)acetonitrile
Molecular Weight (g/mol): 194.24
Molecular Formula: C13H10N2
InChi Key: CAXNYFPECZCGFK-UHFFFAOYNA-N
SMILES: N#CC(C1=CC=CC=C1)C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results