missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-(2-pyridyl)acetonitrile 98.0+%, TCI America™

Catalog No. P20941G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P20941G Supplier TCI America Supplier No. P20941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5005-36-7
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.237
MDL Number MFCD00023497
InChI Key CAXNYFPECZCGFK-UHFFFAOYSA-N
Synonym alpha-Phenyl-2-pyridineacetonitrile, 2-(alpha-Cyanobenzyl)pyridine
PubChem CID 95510
IUPAC Name 2-phenyl-2-pyridin-2-ylacetonitrile
SMILES C1=CC=C(C=C1)C(C#N)C2=CC=CC=N2

Specifications

Melting Point 88°C
Color White-Yellow
Boiling Point 150°C
Quantity 1 g
UN Number 3439
Formula Weight 194.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenyl-2-(2-pyridyl)acetonitrile