accessibility menu, dialog, popup

UserName

CAS RN 49855-03-0

IUPAC Name: 3-(3-methoxyphenoxy)propanoic acid
Synonyms: 3-(3-methoxyphenoxy)propanoic acid
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: JKTUZDBNDYMNLO-UHFFFAOYSA-N
SMILES: COC1=CC(OCCC(O)=O)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results