accessibility menu, dialog, popup

UserName

ONONClNCl

CAS RN 49845-33-2

IUPAC Name: 2,4-dichloro-5-nitropyrimidine
Synonyms: 2,4-dichloro-5-nitropyrimidine
Molecular Weight (g/mol): 193.97
Molecular Formula: C4HCl2N3O2
InChi Key: INUSQTPGSHFGHM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=C(Cl)N=C1Cl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)