missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-nitropyrimidine 98.0+%, TCI America™

Catalog No. D37215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37215G 5 g
D372125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37215G Supplier TCI America Supplier No. D37215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 49845-33-2
Molecular Formula C4HCl2N3O2
Molecular Weight (g/mol) 193.97
MDL Number MFCD00127867
InChI Key INUSQTPGSHFGHM-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-nitro-pyrimidine, pyrimidine, 2,4-dichloro-5-nitro, 2, 4-dichloro-5-nitropyrimidine, 3-nitro-2,4-dichloropyrimidine, 2,4-dichloro-5-nitro pyrimidine, zlchem 540, pubchem7079, acmc-1ann4, ksc236i7h, 2,4-dichloro5-nitropyrimidine
PubChem CID 521266
IUPAC Name 2,4-dichloro-5-nitropyrimidine
SMILES [O-][N+](=O)C1=CN=C(Cl)N=C1Cl

Specifications

Boiling Point 139°C
Quantity 5 g
Formula Weight 193.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dichloro-5-nitropyrimidine
TSCA No
Recommended Storage Refrigerator