accessibility menu, dialog, popup

UserName

CAS RN 4984-85-4

IUPAC Name: 4-hydroxyhexan-3-one
Synonyms: 4-hydroxy-3-hexanone propioin
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: SKCYVGUCBRYGTE-UHFFFAOYNA-N
SMILES: CCC(O)C(=O)CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results