accessibility menu, dialog, popup

UserName

CAS RN 498-15-7

IUPAC Name: (1S,6R)-3,7,7-trimethylbicyclo[4.1.0]hept-3-ene
Synonyms: (+)-delta(3)-carene (+)-delta3-carene 1α,6α-car-3-ene isodiprene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: BQOFWKZOCNGFEC-BDAKNGLRSA-N
SMILES: CC1=CC[C@@H]2[C@H](C1)C2(C)C
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)