missing translation for 'onlineSavingsMsg'
Learn More

(+)-3-Carene 90.0+%, TCI America™

Catalog No. C0047500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0047500ML 500 mL
C004725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0047500ML Supplier TCI America Supplier No. C0047500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 498-15-7
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00066417
InChI Key BQOFWKZOCNGFEC-BDAKNGLRSA-N
PubChem CID 443156
ChEBI CHEBI:7
IUPAC Name (1S,6R)-3,7,7-trimethylbicyclo[4.1.0]hept-3-ene
SMILES CC1=CC[C@@H]2[C@H](C1)C2(C)C

Specifications

Color Yellow
Boiling Point 170°C
Quantity 500 mL
UN Number 2319
Formula Weight 136.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material (+)-3-Carene
EINECSNumber (4)-0589
RTECSNumber FH8400000
TSCA Yes