missing translation for 'onlineSavingsMsg'
Learn More

(+)-3-Carene

Catalog No. AC463990250
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463990250 25 mL
AC463990050 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463990250 Supplier Thermo Scientific Chemicals Supplier No. 463990250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 498-15-7 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 498-15-7
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00066417
InChI Key BQOFWKZOCNGFEC-BDAKNGLRSA-N
IUPAC Name (1S,6R)-3,7,7-trimethylbicyclo[4.1.0]hept-3-ene
SMILES CC1=CC[C@@H]2[C@H](C1)C2(C)C

Specifications

Density 0.8650 g/mL
Boiling Point 170.0°C
Flash Point 46°C
Quantity 25 mL
Specific Gravity 0.865
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in benzene, acetone, ethanol
Formula Weight 136.23
Chemical Name or Material (+)-3-Carene
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Flammable liquid and vapor.

May be fatal if swallowed and enters airways.

Causes skin irritation.

May cause an allergic skin reaction.

Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

Do NOT induce vomiting.

Wear protective gloves/protective clothing.

Wash face, hands and any exposed skin thoroughly after handling.

If skin irritation or rash occurs: Get medical advice/attention.

EINECSNumber 207-856-6

RUO – Research Use Only