missing translation for 'onlineSavingsMsg'
Learn More

(+)-3-Carene, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111018915
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018915 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018915 Supplier Merck Emd Millipore Supplier No. 219865ML

3-Carene is a bicyclic monoterpene, occurring naturally in pine trees.

Featured Industries: Food and Beverages

Specifications

CAS 498-15-7
Density 0.864 g/mL (at 20°C)
Flash Point 47°C - Closed Cup
Physical Form Neat
Linear Formula C10H16
MDL Number MFCD00066417
Boiling Point 170°C to 172°C
Synonym (1 S)-3,7,7-Trimethylbicyclo[4.1.0 ]hept-3-ene
Type Analytical Standard
Quantity 5 mL
Formula Weight 136.23
Percent Purity ≥98.5% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less