accessibility menu, dialog, popup

UserName

CAS RN 4957-14-6

CH3H3C

CAS RN 4957-14-6

IUPAC Name: 1-methyl-4-[(4-methylphenyl)methyl]benzene
Synonyms: 1-methyl-4-[(4-methylphenyl)methyl]benzene
Molecular Weight (g/mol): 196.29
Molecular Formula: C15H16
InChi Key: HZAWPPRBCALFRN-UHFFFAOYSA-N
SMILES: CC1=CC=C(CC2=CC=C(C)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results